Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Formate Solution, 10mM NaOH in isopropanol/water, Solstice

Catalog No. 60047801 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-801 100 mL Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-801 Supplier Solstice Supplier No. 97574100ML

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

suitable for LC-MS, 10 mM NaOH in isopropanol/water 1:1 (+0.2% HCOOH)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Quantity 100 mL
Linear Formula HCOONa
UN Number NONH for all modes of transport
Beilstein 3723428
Formula Weight 68.01g/mol
Packaging Plastic bottle
Chemical Name or Material Sodium formate solution
Hazard Statement P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.