Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexafluorophosphate, 99+%

Catalog No. AA1306422
Click to view available options
Quantity:
25 g
100 g

CAS: 21324-39-0 | F6NaP | 167.95 g/mol

Sodium hexafluorophosphate is used as an ion pairing agent for the determination of cobalt(II) in water samples. It is used as an important raw material in the preparation of 2-chloro-1,3-bis(dimethylamino)trimethinium hexafluorophosphate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21324-39-0
Molecular Formula F6NaP
Molecular Weight (g/mol) 167.95
MDL Number MFCD00011122
InChI Key KMADQUOCJBLXRP-UHFFFAOYSA-N
Synonym sodium hexafluorophosphate, sodium hexa-fluoro phosphate, sodium hexafluorophosphate v, napf6, sodium hexaflurophosphate, acmc-1cao2, sodium hexafluoro-, e5-phosphanuide, sodium hexafluoro-$l^ 5-phosphanuide, phosphate 1-, hexafluoro-, sodium 1:1
PubChem CID 5147921
IUPAC Name sodium;hexafluorophosphate
SMILES [Na+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point >200°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range >99%
Solubility Information Soluble in water.
Formula Weight 167.95
Odor Odorless
Sensitivity Hygroscopic
Appearance White crystalline
Density 2.37
Chemical Name or Material Sodium hexafluorophosphate
Show More Show Less
Hazard Category H302+H312+H332-H314
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8, Packing Group: II
EINECSNumber 244-333-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.