Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydrogen carbonate, 99%

Catalog No. AAA170050C
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA170050C 10,000 g
AAA1700536 500 g
AAA170050E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA170050C Supplier Thermo Scientific Chemicals Supplier No. A170050C
Add to Cart

CAS: 144-55-8 | CHNaO3 | 84.01 g/mol

Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium hydrogen carbonate is used as a laboratory chemical, a leavening agent, a neutralizer, as an active component in fire extinguishers, as a source of carbon dioxide and pH balancer in pools, as a cattle feed supplement and in spas and garden ponds. It is utilized for paint and corrosion removal, baking powder, as a mild abrasive and in cleaning compounds. In the field of medicine, it is used as an antacid to treat acid indigestion and heartburn.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong acids and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Formula CHNaO3
Molecular Weight (g/mol) 84.01
MDL Number MFCD00003528
InChI Key UIIMBOGNXHQVGW-UHFFFAOYSA-M
Synonym sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut
PubChem CID 516892
ChEBI CHEBI:32139
IUPAC Name sodium hydrogen carbonate
SMILES [Na+].OC([O-])=O

Specifications

pH 8.1 to 8.4
Quantity 10,000 g
Beilstein 4153970
Merck Index 14,8583
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
Formula Weight 84.01
Percent Purity 99%
Odor Odorless
Refractive Index 1.5
Density 2.159
Chemical Name or Material Sodium hydrogen carbonate
Show More Show Less
EINECSNumber 205-633-8
RTECSNumber VZ0950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.