Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydrogen Carbonate, MilliporeSigma

Catalog No. 01213171 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
12 kg
25 kg
50 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-213-171 50 kg
01-213-172 500 g
01-213-170 1 kg
01-213-169 5 kg
01-213-174 12 kg
01-213-175 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-213-171 Supplier MilliporeSigma Supelco Supplier No. 1063299050
Add to Cart
Add to Cart

Sodium hydrogen carbonate for analysis EMSURE™ ACS,Reag.Ph Eur

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-55-8
Molecular Weight (g/mol) 84.01

Specifications

Physical Form Powder
Quantity 50 kg
Percent Purity Not Measured
Grade ACS
Packaging Fiber Carton
Chemical Name or Material Sodium hydrogen carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.