Learn More
Strem, An Ascensus Company Sodium bisulfide anhydrous 25g
SDP
Supplier:  Strem, An Ascensus Company 93106125G
Encompass_Preferred
CAS# 16721-80-5. 25g. Sodium hydrogen sulfide, anhydrous. MFCD00011124. Molecular Weight: 56.06. Molecular Formula: NaHS. Color/form: off-white pwdr. Strem# 93-1061. http://www.strem.com/catalog/v/93-1061/
Catalog No. 50-901-13810
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.