Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.0200 N (N/50) in Methanol, Ricca Chemical
SDP

Catalog No. 74710216 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
74710216 500 mL Natural Poly Bottle
74710232 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74710216 Supplier Ricca Chemical Company Supplier No. 7471.0216
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 67-56-1 , 7732-18-5
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
IUPAC Name methanol
SMILES CO

Specifications

Color Colorless
CAS Min % 0.19, 95.0, 2.35
CAS Max % 0.2, 99.0, 2.44
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Methanol
pH >12
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 0.0200Normal
Chemical Name or Material Sodium Hydroxide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.