Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.100 N Solution, APHA, Spectrum™ Chemical
SDP

Catalog No. 18606526 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
4 x 4 L
6 x 1 L
12 x 1 L
200 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-606-526 12 x 1 L
18-606-529 500 mL
18-606-525 4 L
18-606-530 6 x 1 L
18-606-527 4 x 4 L
18-606-528 20 L
18-606-531 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-606-526 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S3401LTCS12
Add to Cart
Add to Cart

S-340, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 0.100 N Solution, APHA is a standard solution of Caustic soda ideally suited for laboratory titrimetric techniques. You can rely on the quality of LabChem analytical test solutions for lot-to-lot reproducibility.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.004, 0.996
Concentration 0.1 N ± 0.003 N
Quantity 12 x 1 L
Grade APHA
Packaging Plastic Bottle
Chemical Name or Material Sodium Hydroxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.