Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.100 Normal (N/10), Reagents

Catalog No. 01336299
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Poly Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-336-299 20 L Cubitainer
01-336-303 100 mL Poly Bottle
01-336-302 10 L Cubitainer
01-336-301 1 L Poly Bottle
01-336-298 4 L Poly Bottle
01-336-296 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-336-299 Supplier Reagents Holdings Llc Supplier No. C731800020F
Add to Cart
Add to Cart

Sodium Hydroxide, 0.100 Normal (N/10), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH 13
Color Colorless
Boiling Point 100°C
Quantity 20 L
Grade Reagent
Packaging Cubitainer
Specific Gravity 1.0046
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 0.100 Normal (N/10)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.