Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.100 N (N/10) in Denatured Alcohol, Ricca Chemical
SDP

Catalog No. 747732 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
747732 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 747732 Supplier Ricca Chemical Company Supplier No. 747732
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory grade, 0.100 N sodium hydroxide solution in denatured alcohol.

Chemical Identifiers

CAS 64-17-5 , 67-56-1 , 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
MDL Number MFCD00004595
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Solubility Information Solubility in Water: Infinite
Quantity 1 L
Grade Laboratory
Odor Alcohol
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution
Recommended Storage Flammable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.