Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.500 N (N/2), Ricca Chemical
SDP

Catalog No. 74001CT Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
74001CT 4 L Cubitainer
740016 500 mL Natural Poly Bottle
7400-32 1 L Natural Poly Bottle
7400-1 4 L Natural Poly Bottle
74002-5 10 L Cubitainer
7400-5 20 L Cubitainer
RIC01003ST 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 74001CT Supplier Ricca Chemical Company Supplier No. 74001CT
Add to Cart
Add to Cart
  • Tested in an ISO 17025 Accredited Facility
  • Test Reports provided with Certified Values and Measurement Uncertainty
  • Tight release specifications for low lot-to-lot variability

Chemical Identifiers

Molecular Formula HNaO
Molecular Weight (g/mol) 40 g/mol
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
CAS Min % 94
CAS Max % 98
Concentration or Composition Notes 0.500 N
Quantity 4 L
Grade Laboratory
Packaging Cubitainer
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.