Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 1.20 N, Ricca Chemical
SDP

Catalog No. R745020015E Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
20 L
15 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R745020015E 15 gal. Poly Drum
R74502001A 1 L Natural Poly Bottle
R745020020F 20 L Cubitainer
R745020020E 20 L Hard Pack
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. R745020015E missing translation for 'mfr' Ricca Chemical Company missing translation for 'supplierNo' R745020015E
Add to Cart
Add to Cart

Standardized Bases

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

pH >13
Color Colorless
Boiling Point 100°C
Melting Point 0°C
CAS Min % 95.42, 4.58
CAS Max % 95.42, 4.58
Concentration 1.199 to 1.201 N
Density 1.051
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Solubility Information Miscible in water
Quantity 15 gal.
IUPAC Name sodium;hydroxide
Molecular Weight (g/mol) 39.997
Grade Laboratory
Packaging Poly Drum
Physical Form Liquid
Assay Normal at 20°C
Chemical Name or Material Sodium Hydroxide, 1.20 Normal
Show More Show Less
ShelfLife 24 months
Recommended Storage Ambient Corrosive
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.