Learn More
Specifications
Specifications
| Color | Colorless |
| Concentration | 2.499 to 2.501 N |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| Quantity | 1 L |
| IUPAC Name | sodium;hydroxide |
| Molecular Weight (g/mol) | 39.997 |
| Grade | Laboratory |
| Concentration or Composition (by Analyte or Components) | 2.50 N |
| Packaging | Natural Poly Bottle |
| Physical Form | Liquid |
| Show More |
Safety and Handling
| ShelfLife | 24 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
