Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 25% (w/w), Analytical Reagent Grade, Ricca Chemical
SDP

Catalog No. R727700020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
20 L
Packaging:
Cubitainer
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R727700020F 20 L Cubitainer
R7277000500 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. R727700020F Supplier Ricca Chemical Company Supplier No. 72775
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Assay Percent Range 25.0 ± 0.1% (w/w)
Quantity 20 L
Percent Purity 24.9 to 25.1%
Grade Analytical
Odor Odorless
Packaging Cubitainer
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 25% (w/w) Aqueous Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.