Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 40% (w/v), Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis, Ricca Chemical
SDP

Catalog No. R72800001A Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
Packaging:
Cubitainer
Natural Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
R72800001A 1 L Natural Poly Bottle
R7280000500 500 mL Natural Poly Bottle
7280-1 4 L Natural Poly Bottle
R728000010F 10 L Cubitainer
7280-5 20 L Cubitainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. R72800001A Supplier Ricca Chemical Company Supplier No. R72800001A
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Color Colorless
Synonym sodium hydroxide
Quantity 1 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity 39.8 to 40.2%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide Solution, 40% (w/v), Nitrogen Free
Show More Show Less
ShelfLife 36 months
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.