Learn More
Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Color | Colorless |
| CAS Min % | 49.0 |
| CAS Max % | 51.0 |
| Concentration or Composition Notes | 50% (w/w), 760 g/L |
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| Quantity | 15 g |
| IUPAC Name | sodium;hydroxide |
| Molecular Weight (g/mol) | 39.997 |
| Percent Purity | min 49.5% w/w |
| Grade | Purified |
| Show More |
Safety and Handling
| ShelfLife | 36 months |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.