Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical
SDP

Catalog No. 72905 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
15 g
500 mL
1 L
20 L
55 gal.
Packaging:
Cubitainer
Hard Pack
Natural Poly Bottle
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
7290-5 20 L Cubitainer
729016 500 mL Natural Poly Bottle
729015G 15 g Poly Drum
729032 1 L Natural Poly Bottle
7290-5HP 20 L Hard Pack
R729000055E 55 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 7290-5 Supplier Ricca Chemical Company Supplier No. 72905
Add to Cart
Add to Cart

Nitrogen Free - Suitable for Kjeldahl Nitrogen Analysis

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
PubChem CID 14798
ChEBI CHEBI:32145
SMILES [OH-].[Na+]

Specifications

Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
Quantity 20 L
IUPAC Name sodium hydroxide
Molecular Weight (g/mol) 40.00
Percent Purity Min. 49.5%
Grade Purified
Concentration or Composition (by Analyte or Components) 50% (w/w) Aqueous Solution (760 g/L)
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide Solution, Nitrogen Free
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.