Learn More
Sodium Hydroxide, 50% (w/w) (760 g/L), Purified Grade, Ricca Chemical
Shop All Ricca Chemical Company Products

Specifications
Specifications
| Synonym | sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic |
| Quantity | 20 L |
| IUPAC Name | sodium hydroxide |
| Molecular Weight (g/mol) | 40.00 |
| Percent Purity | Min. 49.5% |
| Grade | Purified |
| Concentration or Composition (by Analyte or Components) | 50% (w/w) Aqueous Solution (760 g/L) |
| Packaging | Hard Pack |
| Chemical Name or Material | Sodium Hydroxide Solution, Nitrogen Free |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.