Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 96+% Caustic Soda Beads, Reagents

Catalog No. 01337010
Change view
Click to view available options
Quantity:
500 g
3 kg
10 lb.
10 kg
50 lb.
50 kg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-010 50 lb.
01-337-016 10 lb.
01-337-015 10 kg
01-337-013 3 kg
01-337-012 500 g
01-337-011 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-337-010 Supplier Reagents Holdings Llc Supplier No. C231825050F5
Add to Cart
Add to Cart

Sodium Hydroxide, 96+% Caustic Soda Beads, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym Lye, Caustic soda
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 50 lb.
Grade Pure
Packaging Fiber Drum
Specific Gravity 2.13
Physical Form Solid
Chemical Name or Material Sodium Hydroxide, 96+% Caustic Soda Beads
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.