Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, ACS Reagent (Pellets), Reagents

Catalog No. 01337727
Change view
Click to view available options
Quantity:
500 g
1 kg
3 kg
10 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-727 3 kg
01-337-729 10 kg
01-337-728 1 kg
01-337-726 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 01-337-727 Supplier Reagents Holdings Llc Supplier No. C13180003A5
Add to Cart
Add to Cart

Sodium Hydroxide, ACS Reagent (Pellets), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym Lye, Caustic soda
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

pH 14
Color White
Boiling Point 1390°C
Melting Point 318°C
Quantity 3 kg
Grade ACS
Packaging Poly Bottle
Specific Gravity 2.13
Physical Form Solid
Chemical Name or Material Sodium Hydroxide, ACS Reagent (Pellets)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.