Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide Monohydrate, Solstice

Catalog No. 6001398 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-013-98 100 g
60-013-99 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-013-98 Supplier Solstice Supplier No. 01968100G
Add to Cart
Add to Cart

TraceSELECT™, for trace analysis,≥99.9995% (metals basis)

TraceSELECT™ Ultra and TraceSELECT

  • ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
  • sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
  • the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
  • packaged in high-quality containers appropriate for each product
  • the reagents are produced and bottled under clean-room conditions
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12200-64-5
Molecular Formula H3NaO2
Molecular Weight (g/mol) 58.012
MDL Number MFCD00149229
InChI Key JBJWASZNUJCEKT-UHFFFAOYSA-M
Synonym sodium hydroxide monohydrate, na.ho.h2o, naoh water, water naoh, sodium hydroxide h2o, sodium hydroxide water, water sodium hydroxide, h2o naoh, naoh h2o, sodium,hydroxide,hydrate
PubChem CID 23719242
IUPAC Name sodium;hydroxide;hydrate
SMILES O.[OH-].[Na+]

Specifications

Linear Formula NaOH · H2O
UN Number 1823
Quantity 100 g
Formula Weight 58.01g/mol
Percent Purity ≥99.99% (metals basis)
Grade TraceSELECT
Chemical Name or Material Sodium Hydroxide Monohydrate
Hazard Statement P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338
Precautionary Statement H314
EINECSNumber 215-185-5
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.