Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, Pellets, NF, 95-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18606581 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-581 6 x 500 g Poly Bottle
18-606-582 2.5 kg Poly Bottle
18-606-583 4 x 2.5 kg Poly Bottle
18-606-584 12 kg Poly Pail
18-606-585 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-606-581 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO170500GMCS6
Add to Cart
Add to Cart

SO170, 1310-73-2, NaOH

Spectrum™ Chemical Sodium Hydroxide, Pellets, NF is used as a pH adjusting agent. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 1
Quantity 6 x 500 g
Percent Purity 95 to 100.5%
Grade NF
Packaging Poly Bottle
Chemical Name or Material Sodium Hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.