Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N), Solstice

Catalog No. 6001439 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
2 L
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-014-39 2 L Poly Bottle
60-014-40 500 mL Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-014-39 Supplier Solstice Supplier No. 3195032L
Add to Cart
Add to Cart

Volumetric, 0.5 M NaOH (0.5N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
MDL Number MFCD00003548
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Boiling Point >100°C
CAS Min % ≥90.0000%, ≥1.0000%
CAS Max % ≤100.0000%, <2.0000%
Linear Formula NaOH
Concentration 0.5 M NaOH (0.5N)
UN Number UN1824
Density 1.02 g/mL at 25 °C
Quantity 2 L
Formula Weight 40.00
Grade Volumetric
Packaging Poly Bottle
Chemical Name or Material Sodium hydroxide solution, Volumetric, 0.5 M NaOH (0.5N)
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Sodium hydroxide solution
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.