Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hypochlorite, Solution, 12% Available Chlorine, Reagent, Spectrum™ Chemical
SDP

Catalog No. 18606592 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
6 x 500 mL
4 x 4 L
200 L
Packaging:
Drum
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-592 6 x 500 mL Poly Bottle
18-606-594 4 x 4 L Poly Bottle
18-606-595 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-592 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1316500MLPLCS6
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

S1316, 7681-52-9

Spectrum™ Chemical Sodium Hypochlorite, Solution, 12 Percent Available Chlorine, Reagent is made from passing chlorine gas through cold and dilute sodium hydroxide solution. It is most commonly used as a bleaching agent to remove stains from laundry in the household. Sodium hypochlorite is also used for disinfection and cleaning.

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.44
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
IUPAC Name sodium hypochlorite
SMILES [Na+].[O-]Cl

Specifications

CAS Max % 0.13, 0.87
Quantity 6 x 500 mL
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.