Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hypochlorite solution, 6-14% active chlorine basis, Solstice

Catalog No. 60044803 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
6 x 500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-803 6 x 500 mL
60-014-86 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-803 Supplier Solstice Supplier No. 134406X500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

6-14% active chlorine basis

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9 , 7732-18-5
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Boiling Point 111°C
CAS Min % ≥70.0000%, ≥10.0000%
CAS Max % <90.0000%, <20.0000%
Quantity 6 x 500 mL
Linear Formula NaClO
UN Number UN1791
Formula Weight 74.44
Percent Purity 6 to 14% (active chlorine basis)
Packaging Poly Bottle
Density 1.206 g/cm3
Chemical Name or Material Sodium hypochlorite solution, 6-14% active chlorine basis
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P310
Precautionary Statement H314-H400
ShelfLife 240 days from date of manufacture
DOTInformation Transport Hazard Class: 8; Packing Group:3: III; Proper Shipping Name: Hypochlorite solutions
Recommended Storage 2° to 8°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.