Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hypochlorite Solution, 6% (v/v), Ricca Chemical
SDP

Catalog No. 7495632 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-956-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 74-956-32 Supplier Ricca Chemical Company Supplier No. 7495.632
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7732-18-5 , 7681-52-9
Molecular Weight (g/mol) 74.44 g/mol
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

Physical Form Solution
Quantity 1 L
Assay Percent Range 6% (v/v)
Solubility Information Solubility in Water: Infinite
Percent Purity 6% V/V
Grade Laboratory
Odor Mild Chlorine odor
Packaging Amber Poly Bottle
Appearance Clear colorless to slight yellow liquid
Chemical Name or Material Sodium Hypochlorite
Recommended Storage General
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.