Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hypochlorite solution, purum, ≈10% (RT), Solstice

Catalog No. 6001487 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
10 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-014-87 10 L
60-014-88 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-014-87 Supplier Solstice Supplier No. 7169610L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purum, ∼10% (RT)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7681-52-9
Molecular Formula ClNaO
Molecular Weight (g/mol) 74.439
MDL Number MFCD00011120
InChI Key SUKJFIGYRHOWBL-UHFFFAOYSA-N
Synonym sodium hypochlorite, antiformin, sodium oxychloride, chlorox, clorox, javex, javelle water, hypochlorous acid, sodium salt, hypochlorite sodium, carrel-dakin solution
PubChem CID 23665760
ChEBI CHEBI:32146
IUPAC Name sodium;hypochlorite
SMILES [O-]Cl.[Na+]

Specifications

Physical Form Liquid
Quantity 10 L
Linear Formula NaClO
UN Number UN1791
Formula Weight 74.44g/mol
Percent Purity 10% (RT)
Grade purum
Concentration or Composition (by Analyte or Components) ≈10% (RT)
Density 1.21-1.26g/mL at 20°C
Chemical Name or Material Sodium hypochlorite solution
Hazard Statement P273-P280-P303 + P361 + P353-P304 + P340 + P310-P305 + P351 + P338-P391
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.