Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Iodide, 1M
SCI_ED

Catalog No. S25555 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
1M
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25555 500 mL 1M
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25555

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7681-82-5
Molecular Formula INa
Molecular Weight (g/mol) 149.894
InChI Key FVAUCKIRQBBSSJ-UHFFFAOYSA-M
Synonym sodium iodide, ioduril, sodium monoiodide, soiodin, sodiumiodide, natrii iodidum, jodid sodny, iodure de sodium, jodid sodny czech
PubChem CID 5238
ChEBI CHEBI:33167
IUPAC Name sodium;iodide
SMILES [Na+].[I-]

Specifications

Quantity 500 mL
Assay Percent Range >99%
Linear Formula NaI
Formula Weight 149.89
Packaging 1M
Density 3.6670g/cmsuperscript_3;
Chemical Name or Material Sodium Iodide
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.