Learn More
Description
- Material: Sodium Iodide (NaI)
- Color: White
- Quantity: 500 grams
- Assay Percent Range: Greater than 99% purity
- Packaging: Lab Grade Powder
- Density: 3.667 g/cm³
- Usage: For educational purposes only, no Certificate of Analysis (C of A) available
- Applications: Ideal for use in chemistry experiments, radiation detection studies, and physics demonstrations
This product is for educational use - no C of A is available.
Specifications
Specifications
| Color | White |
| Physical Form | Powder, Crystals and/or Chunks |
| Quantity | 500 g |
| Assay Percent Range | >99% |
| Linear Formula | NaI |
| Formula Weight | 149.89 |
| Packaging | Lab Grade |
| Density | 3.6670g/cmsuperscript_3; |
| Chemical Name or Material | Sodium Iodide |
Safety and Handling
| ShelfLife | 1 year |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
