Sodium iodide, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1166522

Quantity 100g

 View more versions of this product

Catalog No. AA1166522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7681-82-5
    Physical Form Powder
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula NaI
    Formula Weight 149.89
    MDL Number MFCD00003532
    Chemical Name or Material Sodium iodide

    Specifications

    Formula Weight 149.89
    Quantity 100g
    Color White
    EINECS Number 231-679-3
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula NaI
    Appearance White powder or beads
    MDL Number MFCD00003532
    CAS 7681-82-5
    Health Hazard 1 H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Sodium iodide

    H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit