Learn More
Sodium iodide, ultra dry, 99.98% (metals basis)
Description
It is used as iodine supplement, expectorant or in its radioactive (I-131) form as an diagnostic aid, particularly for thyroid function tests. sodium Iodide are used in photography and as analytical reagents. It is also used in the measurement of the energy of gamma rays, also used in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Boiling Point | 1300°C to 1304°C |
| Melting Point | 651°C |
| Physical Form | Beads |
| Quantity | 500 g |
| Packaging | Ampouled under argon |
| Refractive Index | 1.7745 |
| Sensitivity | Hygroscopic |
| Specific Gravity | 3.67 |
| Density | 3.67 |
| Solubility Information | It is soluble in water,alcohol,and glycerin. |
| Show More |
Safety and Handling
| Hazard Category | H315-H319 |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| DOTInformation | Hazard Class: 9; Packaging Group: III |
| EINECSNumber | 231-679-3 |
| RTECSNumber | WB6475000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.