Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Isonicotinate 99.0+%, TCI America™
SDP

Catalog No. P0991250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0991250G 250 g
P099125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P0991250G Supplier TCI America Supplier No. P0991250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16887-79-9
Molecular Formula C6H4NNaO2
Molecular Weight (g/mol) 145.093
MDL Number MFCD00064868
InChI Key PVXCGWNPDFFPOV-UHFFFAOYSA-M
Synonym Isonicotinic Acid Sodium Salt, Pyridine-4-carboxylic Acid Sodium Salt, Sodium Pyridine-4-carboxylate
PubChem CID 23662338
IUPAC Name sodium;pyridine-4-carboxylate
SMILES C1=CN=CC=C1C(=O)[O-].[Na+]

Specifications

Color White
pH 7.0 to 10.0 (50g/L H2O 25°C)
Quantity 250 g
Formula Weight 145.09
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Isonicotinate
RTECSNumber NS2100000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.