Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metabisulfite, Granular, NF, EP, BP, JP, 65-67.4%, Spectrum™ Chemical
SDP

Catalog No. 18606654 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-654 12 kg Poly Pail
18-606-653 2.5 kg Poly Bottle
18-606-655 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-654 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. SO18212KG
Add to Cart
Add to Cart

SO182, 7681-57-4, Na2S2O5

Spectrum™ Chemical Sodium Metabisulfite, Granular, NF, EP, BP, JP is commonly used as a preserving agent, antioxidant and disinfectant. All Spectrum Chemical NF, EP, BP, and JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
MDL Number MFCD00167602
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

CAS Max % 1
Quantity 12 kg
Percent Purity 65 to 67.4%
Grade Multi-Compendial
Packaging Poly Pail
Appearance Clear
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.