Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Metabisulfite, Purified *This material is a mixture containing some Sodium Metabisulfite (CAS# 7681-57-4), Reagents

Catalog No. 01337002
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-002 500 g
01-337-003 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-002 Supplier Reagents Holdings Llc Supplier No. C2320100500A5
Add to Cart
Add to Cart

Sodium Metabisulfite, Purified *This material is a mixture containing some Sodium Metabisulfite (CAS# 7681-57-4), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7681-57-4
Molecular Formula Na2O5S2
Molecular Weight (g/mol) 190.09
InChI Key LDTLADDKFLAYJA-UHFFFAOYSA-L
Synonym Disodium Disulfite, Sodium Pyrosulfite
IUPAC Name disodium (sulfinatooxy)sulfinate
SMILES [Na+].[Na+].[O-]S(=O)OS([O-])=O

Specifications

Melting Point 150°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.48
Chemical Name or Material Sodium Metabisulfite, Purified *This material is a mixture containing some Sodium Metabisulfite (CAS# 7681-57-4)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.