Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Methoxide, 0.010 Molar (Sodium Methylate, 0.010 Normal), Reagents

Catalog No. 01333801
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-801 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-801 Supplier Reagents Holdings Llc Supplier No. CS1089001C
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Methoxide, 0.010 Molar (Sodium Methylate, 0.010 Normal), Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 67-56-1 , 124-41-4
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Sodium Methylate, Sodium Methanolate, Methoxysodium
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point 65°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 1 L
Grade Reagent
Packaging Glass Bottle
Specific Gravity 0.8
Chemical Name or Material Sodium Methoxide, 0.010 Molar (Sodium Methylate, 0.010 Normal)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.