Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium methoxide, pure, titrant, (0.5 M in methanol)

Catalog No. ac419605000
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC419605000 500 mL Glass bottle
AC419601000 100 mL Glass bottle
AC419600025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. AC419605000 Supplier Thermo Scientific Chemicals Supplier No. 419605000
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 124-41-4 | CH3NaO | 54.024 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-41-4 , 67-56-1
Molecular Formula CH3NaO
Molecular Weight (g/mol) 54.024
MDL Number MFCD00012179
InChI Key WQDUMFSSJAZKTM-UHFFFAOYSA-N
Synonym sodium methanolate, sodium methoxide, sodium methylate, methoxysodium, methanol, sodium salt, feldalat nm, metilato sodico spanish, unii-ig663u5emc, methylate de sodium french, hsdb 755
PubChem CID 10942334
IUPAC Name sodium;methanolate
SMILES C[O-].[Na+]

Specifications

Color Colorless
Boiling Point 65.0°C
Melting Point -98.0°C
Linear Formula CH3ONa
Concentration 0.45 to 0.60M
Fieser 01,1091; 02,385; 03,259; 04,457; 05,617; 06,545; 08,463; 11,489; 13,352
Density 0.8000g/mL
Solubility Information Solubility in water: reacts.
Formula Weight 54.02
Grade Pure
Packaging Glass bottle
Specific Gravity 0.8
Quantity 500 mL
Flash Point 11°C
Physical Form Liquid
Chemical Name or Material Sodium methoxide, 0.5 M in methanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes serious eye irritation. Causes skin irritation. Causes damage to organs. Highly flammable liquid and vapor. Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
EINECSNumber 204-699-5
RTECSNumber PC3570000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.