Learn More
Apexbio Technology LLC Sodium Monofluorophosphate 10163-15-2 10mM (in 1mL H20)
SDP
Supplier:  Apexbio Technology LLC B18375.1
Encompass_Preferred
Sodium Monofluorophosphate is a small-molecule inhibitor targeting enzymes including pyruvate kinase alkaline phosphatase and phosphorylase phosphatase It is designed to competitively and irreversibly inhibit these enzymes thereby modulating enzyme-mediated phosphorylation processes and impacting glycogen metabolism pathways Sodium Monofluorophosphate exerts its biological activity primarily through competitive inhibition of pyruvate kinase and alkaline phosphatase by interfering with endogenous phosphate substrates and irreversible inhibition of phosphorylase phosphatase Based on these pharmacological properties Sodium Monofluorophosphate holds research potential in the study of enzymatic mechanisms involving phosphate-transfer reactions and in investigations of metabolic regulations dependent on pyruvate kinase alkaline phosphatase and phosphorylase phosphatase pathways
Catalog No. 50-000-92267
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.