Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium n-octyl sulfate, 99%

Catalog No. p-7081707
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1178603 1 g
AAA1178606 5 g
AAA1178614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1178603 Supplier Thermo Scientific Chemicals Supplier No. A1178603
Only null left
Add to Cart

CAS: 142-31-4 | C8H17NaO4S | 232.27 g/mol

Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-octyl sulfate is a useful chemical to study the selective enrichment of albumin in biological samples and to determine the mixed solution of a glycerin-modified cationic surfactant and an anionic surfactant. It is also used as wetting agent, detergent, dispersant, emulsifier, degreasing agent, hard surface cleaner, lime soap dispersant, alkaline detergent, cleaning agent for metal processing in electroplating, latex, metal processing, paper, polymer synthesis, textile, leather and other industries. It finds application as an anionic detergent and ion -pairing reagent used in high-performance liquid chromatography (HPLC).

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-31-4
Molecular Formula C8H17NaO4S
Molecular Weight (g/mol) 232.27
MDL Number MFCD00007470
InChI Key WFRKJMRGXGWHBM-UHFFFAOYSA-M
Synonym sodium octyl sulfate, sodium n-octyl sulfate, sodium octyl sulphate, sipex ols, cycloryl os, duponol 80, sodium capryl sulfate, octyl sodium sulfate, sulfuric acid, monooctyl ester, sodium salt, octyl sulfate sodium salt
PubChem CID 2735107
IUPAC Name sodium;octyl sulfate
SMILES CCCCCCCCOS(=O)(=O)[O-].[Na+]

Specifications

Melting Point ∼195°C (decomposition)
Quantity 1 g
Beilstein 3658635
Solubility Information Soluble in water.
Formula Weight 232.28
Percent Purity 99%
Chemical Name or Material Sodium n-octyl sulfate
Hazard Category H315-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362
EINECSNumber 205-535-5
RTECSNumber WT1175000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.