Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Sodium Nitrite
SCI_ED

Catalog No. s25560a Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Reagent Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25560A 500 g Reagent Grade
S25560 100 g Reagent Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25560A
Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium nitrite
SMILES [Na+].[O-]N=O

Specifications

Color Yellow
Quantity 500 g
Assay Percent Range >95%
Linear Formula NaNO2
Formula Weight 69
Grade Reagent
Packaging Reagent Grade
Chemical Name or Material Sodium Nitrite
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 5.1 (6.1), III, : Poison
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.