Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium nitrite solution, 0.2M, Solstice

Catalog No. 6002634 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-026-34 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-026-34 Supplier Solstice Supplier No. 352721L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Volumetric, 0.2 M NaNO2 (0.2N)

Solstice's high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use
  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7632-00-0
Molecular Formula NNaO2
Molecular Weight (g/mol) 69.00
MDL Number MFCD00011118
InChI Key LPXPTNMVRIOKMN-UHFFFAOYSA-M
Synonym sodium nitrite, nitrous acid, sodium salt, sodiumnitrite, erinitrit, filmerine, natrium nitrit, nitrito sodico, anti-rust, nitrite, sodium, diazotizing salts
PubChem CID 23668193
ChEBI CHEBI:78870
IUPAC Name sodium;nitrite
SMILES [Na+].[O-]N=O

Specifications

Physical Form Liquid
Quantity 1 L
Linear Formula NaNO2
UN Number NONH for all modes of transport
Formula Weight 69g/mol
Grade Not Available
Concentration or Composition (by Analyte or Components) 0.2M
Chemical Name or Material Sodium nitrite solution
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.