Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium nitroferricyanide dihydrate, 99+%

Catalog No. AC211645000
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC211645000 500 g Glass bottle
AC211640250 25 g Glass bottle
AC211641000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC211645000 Supplier Thermo Scientific Chemicals Supplier No. 211645000
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13755-38-9 ∣ C5H4FeN6Na2O3 ∣ 297.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13755-38-9
Molecular Formula C5H4FeN6Na2O3
Molecular Weight (g/mol) 297.95
MDL Number MFCD00149192
InChI Key JFPDVRGGFBUORC-UHFFFAOYSA-N
Synonym nitroprusside sodium, sodium pentacyanonitrosylferrate, sodium nitroferricyanide iii dihydrate, dsstox_cid_21126, dsstox_rid_79630, dsstox_gsid_41126, sodium pentacyanonitrosylferrate iii dihydrate, nitropress tn, sodium nitroprusside usp, c5fen6o.2na.2h2o
PubChem CID 11953895
SMILES O.O.[Na+].[Na+].N#C[Fe++](C#N)(C#N)(C#N)(C#N)[N]#[O+]

Specifications

Color Red
Physical Form Crystals
Quantity 500 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula Na2Fe(CN)5NO·2H2O
Merck Index 15, 8780
Solubility Information Solubility in water: 400g/L (20°C). Other solubilities: slightly soluble in alcohol
IUPAC Name disodium pentacyano(oxidaniumylidyneamino)ironbis(ylium) dihydrate
Formula Weight 297.94
Percent Purity ≥99%
Grade Ungraded
Packaging Glass bottle
Specific Gravity 1.72
Density 1.7200g/mL
Chemical Name or Material Sodium nitroferricyanide dihydrate, p.a.
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
RTECSNumber LJ8925000

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.