Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Oleate, Spectrum™ Chemical
SDP

Catalog No. 18621456 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
50 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-456 500 g
18-621-454 25 g
18-621-455 100 g
18-621-458 50 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-621-456 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1605500GM
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143-19-1
Molecular Weight (g/mol) 304.44 g/mol

Specifications

Quantity 500 g
Physical Form Powder or Waxy Solid
Chemical Name or Material Sodium Oleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.