Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium orthovanadate, 99.9% (metals basis)

Catalog No. AA8110422
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8110422 100 g
AA8110414 25 g
AA8110436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA8110422 Supplier Thermo Scientific Chemicals Supplier No. 08110422
Add to Cart

inhibitor of Protein Tyrosine Phosphatase and Alkaline Phosphate | CAS: 13721-39-6 | Na3O4V | 183.907 g/mol

Inhibitor of protein tyrosine phosphatase and alkaline phosphate Sodium orthovanadate is used in protein analysis in molecular biology and acts as an inhibitor of protein tyrosine phosphatases, alkaline phosphatases and kinases. It is also used to prepare the stop solution in dephosphorylation assay of insulin receptor kinase. It serves as a reagent in the development of matrix Chip that utilizes surface-immobilized antibodies. Further, it is utilized for the preservation of protein tyrosyl phosphorylation state in cells and cell lysates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13721-39-6
Molecular Formula Na3O4V
Molecular Weight (g/mol) 183.907
MDL Number MFCD00003511
InChI Key IHIXIJGXTJIKRB-UHFFFAOYSA-N
Synonym sodium orthovanadate, trisodium vanadate, sodium o-vanadate, sodium pervanadate, sodium vanadate ortho, trisodium tetraoxovanadate, sodium tetraoxovanadate 3-, vanadic acid, trisodium salt, trisodium orthovanadate, vanadic acid h3vo4 , sodium salt
PubChem CID 61671
ChEBI CHEBI:35607
IUPAC Name trisodium;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 850°C to 866°C
Color White
Physical Form Granules
Quantity 100 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Soluble in water.
Formula Weight 183.94
Odor Odorless
Chemical Name or Material Sodium orthovanadate
Hazard Category H302-H315-H319-H335
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 237-287-9
RTECSNumber YW1120000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.