Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Sodium Oxamate, Lactate Dehydrogenase Inhibitor, Solid, 1 g

Catalog No. 502705084
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-5084 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-5084 Supplier Abcam Supplier No. AB1456431G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity sodium oxamate, a weak lactate dehydrogenase inhibitor that suppresses glycolysis and gluconeogenesis.

  • Molecular weight: 111.03 Da
  • Purity: >99%
  • Form: Solid
  • Inhibitor: Weak lactate dehydrogenase inhibitor (IC50 values are 600 and 4400 μM in H460 and A549 cells respectively)
  • Key biological effects: Inhibits glycolysis and gluconeogenesis, decreases extracellular acidification rate
  • Analog: Pyruvate (ab145036) analog
  • CAS Number: 565-73-1
  • Composition: 100% Sodium oxamate

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at -20°C.
Form Solid
Target Molecule Lactate Dehydrogenase
Buffer 100% sodium oxamate
Shipping Condition Ambient
Formulation 100% Sodium Oxamate
Quantity 1 g
For Use With (Application) Inhibition of lactate dehydrogenase, glycolysis and gluconeogenesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.