Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Pentacyanoaminoferrate, Spectrum™ Chemical
SDP

Catalog No. 18606684 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-606-684 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-606-684 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S19681GM

S1968, 36682-41-4, C5H7FeN7Na2

Spectrum™ Chemical Sodium Pentacyanoaminoferrate

Chemical Identifiers

CAS 36682-41-4

Specifications

CAS Max % 1
Quantity 1 g
UN Number UN3439
Grade Reagent
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.