Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Periodate, 6% w/v, Reagents

Catalog No. 01333900
Change view
Click to view available options
Quantity:
4 L
Packaging:
Glass Bottle
Opaque HDPE
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
01-333-900 4 L Opaque HDPE
01-333-899 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-333-900 Supplier Reagents Holdings Llc Supplier No. CS0420004BP
 Restricted Item
Add to Cart
Add to Cart

Sodium Periodate, 6% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 7790-28-5 , 7664-93-9
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Sodium Tetraoxoiodate(VII)
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 4 L
Percent Purity 6%
Grade Reagent
Packaging Opaque HDPE
Specific Gravity 1.05
Chemical Name or Material Sodium Periodate, 6% w/v
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.