Learn More
Sodium periodate, 99.8+%, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 300.0°C |
| Color | White |
| Physical Form | Crystalline Powder |
| Quantity | 2.5 kg |
| Assay Percent Range | 99.8 to 100.3% (on dried basis) (Iodometry) |
| Linear Formula | NaIO4 |
| UN Number | 3087 |
| Fieser | 01,809; 02,311; 04,373; 05,507; 06,455; 11,492; 13,145; 15,294 |
| Merck Index | 15,8772 |
| Solubility Information | Solubility in water: 80g/L (20°C). Other solubilities: 389g/L (52°C), soluble in h2so4, hno3, acetic acids, insoluble in most common organic solvents |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May cause fire or explosion; strong oxidizer. Causes severe skin burns and eye damage. Causes damage to organs through prolonged or repeated exposure if swallowed. Very toxic to aquatic life. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Take any precaution to avoid mixing with combustibles. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if present |
| EINECSNumber | 232-197-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.