Learn More
Sodium peroxide, 96%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 460.0°C |
| Color | Yellow |
| Physical Form | Granules |
| Quantity | 2.5 kg |
| Assay Percent Range | 96% |
| Linear Formula | Na2O2 |
| Fieser | 06,548; 07,341; 11,492 |
| Merck Index | 15, 8786 |
| Solubility Information | Solubility in water: reacts violently. Other solubilities: forming naoh and h2o2,freely soluble |
| Formula Weight | 77.97 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause fire or explosion; strong oxidizer. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses,if pre |
| EINECSNumber | 215-209-4 |
| TSCA | TSCA |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.