Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium peroxide, 96%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
2.5kg
Packaging:
Plastic bottle
1313-60-6
Na2O2
77.97
MFCD00003497
PFUVRDFDKPNGAV-UHFFFAOYSA-N
sodium peroxide,disodium peroxide,solozone,natriumsuperoxid,sodium dioxide,disodium dioxide,natrium peroxydatum,sodium oxide, per,flocool 180,sodium oxide
14803
disodium;peroxide
[O-][O-].[Na+].[Na+]
1313-60-6
Na2O2
77.97
MFCD00003497
PFUVRDFDKPNGAV-UHFFFAOYSA-N
sodium peroxide,disodium peroxide,solozone,natriumsuperoxid,sodium dioxide,disodium dioxide,natrium peroxydatum,sodium oxide, per,flocool 180,sodium oxide
14803
disodium;peroxide
[O-][O-].[Na+].[Na+]
Assay Percent Range 96%
Linear Formula Na2O2
Fieser 06,548; 07,341; 11,492
Merck Index 15, 8786
Solubility Information Solubility in water: reacts violently. Other solubilities: forming naoh and h2o2,freely soluble
Formula Weight 77.97
Physical Form Granules
Percent Purity 96%
Chloride 50ppm max
Chlorine (Cl) 50ppm max.
Density 2.8000g/mL
Iron (Fe) 50ppm max.
Lead (Pb) 50ppm max.
Packaging Plastic bottle
Solubility Solubility in water: reacts violently. Other solubilities: forming naoh and h2o2, freely soluble
Specific Gravity 2.8
Sulfate 50ppm max.
Color Yellow
Melting Point 460.0°C
Quantity 25g
Chemical Name or Material Sodium peroxide, p.a.
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
May cause fire or explosion; strong oxidizer.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 215-209-4

TSCA : TSCA