Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Peroxide, puriss. p.a. ACS Reagent, Solstice

Catalog No. 6001539 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
6 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-015-39 500 g
60-015-38 1 kg
60-045-956 6 x 1 kg
60-015-37 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-015-39 Supplier Solstice Supplier No. 31445500G
Add to Cart
Add to Cart

Puriss. p.a., ACS Reagent, Reag. ISO,≥95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Quantity 500 g
UN Number 1504
Formula Weight 77.98g/mol
Percent Purity ≥95%
Grade ACS/puriss. p.a.
Chemical Name or Material Sodium Peroxide, Reag. ISO
Hazard Statement P220-P280-P305 + P351 + P338-P310
Precautionary Statement H271-H314
EINECSNumber 215-209-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.