Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Peroxide, purum p.a, ≥95%, Solstice

Catalog No. 60047406 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-406 1 kg Poly bottle
60-047-407 250 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-406 Supplier Solstice Supplier No. 718831KG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

for Wurzschmitt-decomposition, ACS reagent, beads (small),≥95%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-60-6
Molecular Formula Na2O2
Molecular Weight (g/mol) 77.978
MDL Number MFCD00003497
InChI Key PFUVRDFDKPNGAV-UHFFFAOYSA-N
Synonym sodium peroxide, disodium peroxide, solozone, natriumsuperoxid, sodium dioxide, disodium dioxide, natrium peroxydatum, sodium oxide, per, flocool 180, sodium oxide
PubChem CID 14803
IUPAC Name disodium;peroxide
SMILES [O-][O-].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
Physical Form Crystals
Quantity 1 kg
UN Number UN1504
Solubility Information 106.7g/L at 20°C
Formula Weight 77.98g/mol
Percent Purity ≥95%
Packaging Poly bottle
Chemical Name or Material Sodium peroxide
Hazard Statement P220-P280-P305 + P351 + P338-P310
DOTInformation 5.1 / PGI
RTECSNumber 215-209-4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.